Thermo Scientific™ Pierce™ Standard Binding NeutrAvidin Coated Plates
Capture biotinylated antibodies or probes for ELISA and other target-specific assays with pre-blocked NeutrAvidin protein-coated plates.
$265.35 - $576.14
Specifications
Binding Target Biotin
Quantity 5 plates
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. Formulation Price Quantity  
Catalog Number Mfr. No. Formulation Price Quantity  
PI15116 Thermo Scientific™
PI15116
White, 96-well plates, coated with 100uL of NeutrAvidin Protein and blocked with 200uL of SuperBlock Blocking Buffer Pack of 5 Each for $296.35
PI15117 Thermo Scientific™
PI15117
Black, 96-well plates, coated with 100uL of NeutrAvidin Protein and blocked with 200uL of SuperBlock Blocking Buffer Pack of 5 Each for $381.06
PI15128 Thermo Scientific™
PI15128
Clear, 96-well strip plates, coated with 200uL of NeutrAvidin Protein and blocked with 300uL of Blocker BSA Pack of 5 Each for $296.35
PI15129 Thermo Scientific™
PI15129
Clear, 96-well plates, coated with 100uL of NeutrAvidin Protein and blocked with 200uL of SuperBlock Blocking Buffer Pack of 5 Each for $439.30
PI15217 Thermo Scientific™
15217
Black, 96-well plates, coated with 200uL of NeutrAvidin Protein and blocked with 300uL of Blocker BSA Pack of 5 Each for $265.35
PI15400 Thermo Scientific™
PI15400
Clear, 384-well plates, coated with 50uL of NeutrAvidin Protein and blocked with 100uL of SuperBlock Blocking Buffer Pack of 5 Each for $548.10
PI15401 Thermo Scientific™
PI15401
White, 384-well plates, coated with 50uL of NeutrAvidin Protein and blocked with 100uL of SuperBlock Blocking Buffer Pack of 5 Each for $576.14
PI15402 Thermo Scientific™
PI15402
Black, 384-well plates, coated with 50uL of NeutrAvidin Protein and blocked with 100uL of SuperBlock Blocking Buffer Pack of 5 Each for $569.12
Specifications
Description
Thermo Scientific™ Pierce NeutrAvidin Coated Plates are pre-blocked and ready-to-use polystyrene 96-well and 384-well plates for binding biotinylated antibodies or probes for ELISA and other target-specific assays.
Biotin-binding plates are ideal for applications when direct adsorption to polystyrene denatures antibodies or the target molecule. Thermo Scientific NeutrAvidin™ Protein is avidin that has been deglycosylated to prevent nonspecific lectin-binding. NeutrAvidin Protein has a near-neutral pI (6.3), resulting in lower nonspecific interactions in certain applications compared with streptavidin. NeutrAvidin Protein has an additional advantage of lacking the bacterial RYD sequence found on streptavidin, which eliminates nonspecific binding due to the RGD binding domain of adhesion receptors present in a variety of cells. Pierce NeutrAvidin Coated Plates are available in clear, white and black plates for use with colorimetric, chemiluminescent and fluorescent detection methods, respectively.
Highlights:
  • Easy and gentle immobilization of biotin-containing conjugates
  • Lowest nonspecific binding properties of all biotin-binding proteins
  • Thermo Scientific NeutrAvidin Biotin-Binding Protein has no carbohydrate and an isoelectric point of 6.3
  • No denaturing of the protein component of a conjugate upon binding to the plate
  • Ideal for binding small hydrophilic molecules (e.g., peptides) that typically exhibit poor binding directly to polystyrene
  • Pre-blocked with your choice of Thermo Scientific Blocker BSA or SuperBlock Blocking Buffer
  • Available in 96- and 384-well formats
Description & Specifications
Specifications
Biotin
5 plates