Please login to your online account to display your discounted pricing
Thermo Scientific™ Pierce™ SILAC Protein Quantitation Kits
Analyze protein expression by mass spectrometry using stable isotope labeling with amino acids in cell culture (SILAC) using these complete reagent kits.
$965.32 - $965.32
Specifications
Quantity 1 use kit
View More Specs
Products
Catalog Number Mfr. No. Description Price Quantity    
PI89982
thermo scientific™
89982
SILAC Protein Quantitation Kit - RPMI 1640 Each for $965.32
PI89983
thermo scientific™
89983
SILAC Protein Quantitation Kit - DMEM Each for $965.32
Description & Specifications
Specifications
Quantity 1 use kit
Thermo Scientific™ SILAC Protein Quantitation Kits are used for the specific analysis of protein expression by mass spectrometry using stable isotope labeling with amino acids in cell culture (SILAC).
SILAC is a powerful method to identify and quantify relative differential changes in complex protein samples. This approach entails the in vivo metabolic incorporation of heavy 13C- or 15N-labeled amino acids into proteins followed by mass spectrometry (MS) for the accelerated and comprehensive identification, characterization and quantitation of proteins.
Highlights:
  • Efficient – 100% label incorporation into proteins of living cells
  • Reproducible – eliminates intra-experimental variability caused by differential sample preparation
  • Flexible – media deficient in both L-lysine and L-arginine, allowing for more complete proteome coverage through dual amino acid isotope labeling
  • Compatible – label proteins expressed in a wide variety of mammalian cell lines adapted to grow in DMEM, RPMI-1640 or DMEM:F12 medium, including HeLa, 293T, COS7, U2OS, A549, A431, HepG2, NIH 3T3, Jurkat and others
  • Optimized, specialized media – the Expansion DMEM and Low-Osmolarity DMEM are specifically designed and qualified for mouse embryonic stem cell propagation and SILAC protein expression profiling with mass spectrometry
  • High-quality supplements – heavy amino acids with >99% isotope purity; the Stem Cell Screened Dialyzed FBS and Serum Substitute for Mouse Embryonic Stem Cells are tested to ensure that they do not promote stem cell differentiation
Recommended for:
Quantitative analysis of relative changes in protein abundance from different cell treatments; Quantitative analysis of proteins for which there are no antibodies available; Protein expression profiling of normal vs. disease cells; Identification and quantification of hundreds to thousands of proteins in a single experiment