Thermo Scientific™ Pierce™ Secondary Antibody Coated Plates
Perform binding assays when available antibodies are in low quantities or they denature and become inactive upon direct adsorption to polystyrene plates.
$282.69 - $303.25
Specifications
Format 96-Well
Quantity 5 plates
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. Color Surface Coating Price Quantity  
Catalog Number Mfr. No. Color Surface Coating Price Quantity  
PI15134 Thermo Scientific™
PI15134
Clear Goat Anti-Mouse IgG Antibody Pack of 5 Each for $282.69
PI15135 Thermo Scientific™
PI15135
Clear Goat Anti-Rabbit IgG Antibody Pack of 5 Each for $282.69
PI15137 Thermo Scientific™
15137
Black Goat Anti-Rabbit IgG Antibody Pack of 5 Each for $303.25
PI15334 Thermo Scientific™
15334
Black Goat Anti-Mouse IgG Antibody Pack of 5 Each for $303.25
Specifications
Description
Thermo Scientific™ Pierce Antibody Coated Plates are ideal for binding assays when available antibodies are in low quantities or they denature and become inactive upon direct adsorption to polystyrene plates.
These plates are pre-coated with goat anti-mouse (GAM) or goat anti-rabbit (GAR) IgG polyclonal antibodies, allowing assay-specific antibodies to be captured even from impure protein samples. The plates are supplied pre-blocked and ready-to-use. Pierce Secondary Antibody Coated Plates can be used for direct, indirect, competitive or sandwich assays and are available in clear for colorimetric assays, white for chemiluminescent assays and black for fluorescent assays.
Highlights:
  • Prevents antibody denaturation caused by direct binding
  • Unlike Protein A or G, these bind only to target species
  • Antibody-binding capacity is higher than direct adsorption
  • Pre-blocked to reduce nonspecific binding
  • Less expensive than Protein A or G plates
Description & Specifications
Specifications
96-Well
5 plates