Please login to your online account to display your discounted pricing

Thermo Scientific™ Pierce™ Secondary Antibody Coated Plates


Perform binding assays when available antibodies are in low quantities or they denature and become inactive upon direct adsorption to polystyrene plates.

Manufacturer: thermo scientific™  15137

 View more versions of this product

Catalog No. PI15137

  • / Pack of 5 Each
 

Description & Specifications

Specifications

Formulation Black, 96-well plates, coated with 100uL of Goat Anti-Rabbit IgG Antibody and blocked with 200uL of SuperBlock Blocking Buffer
Format 96-Well
Quantity 5 plates
Sufficient For Binding approx. 2pmol rabbit IgG per well
Surface Coating Goat Anti-Rabbit IgG Antibody
Binding Target Rabbit IgG
Color Black
Item Description Goat Anti-Rabbit IgG Coated; Black

Thermo Scientific™ Pierce Antibody Coated Plates are ideal for binding assays when available antibodies are in low quantities or they denature and become inactive upon direct adsorption to polystyrene plates.

These plates are pre-coated with goat anti-mouse (GAM) or goat anti-rabbit (GAR) IgG polyclonal antibodies, allowing assay-specific antibodies to be captured even from impure protein samples. The plates are supplied pre-blocked and ready-to-use. Pierce Secondary Antibody Coated Plates can be used for direct, indirect, competitive or sandwich assays and are available in clear for colorimetric assays, white for chemiluminescent assays and black for fluorescent assays.

Highlights:

  • Prevents antibody denaturation caused by direct binding
  • Unlike Protein A or G, these bind only to target species
  • Antibody-binding capacity is higher than direct adsorption
  • Pre-blocked to reduce nonspecific binding
  • Less expensive than Protein A or G plates