Thermo Scientific™ Pierce™ S-Nitrosylation Western Blot Kit
Perform sensitive detection of protein S-nitrosocysteine post-translational modifications with a complete, easy-to-use kit.
$142.00 - $578.00
Products 2
Catalog Number Mfr. No. Description Price Quantity  
Catalog Number Mfr. No. Description Price Quantity  
PI90106 View Documents Thermo Scientific™
90106
HENS Buffer Each for $142.00
PI90105 View Documents Thermo Scientific™
90105*
Pierce S-Nitrosylation Western Blot Kit Each for $578.00
Description
Description
Perform sensitive detection of protein S-nitrosocysteine post-translational modifications with the complete, easy-to-use Thermo Scientific™ Pierce S-Nitrosylation Western Blot Kit.
Additional Information and Online Ordering »
The Pierce S-Nitrosylation Western Blot Kit contains a cell lysis/reaction buffer, a sulfhydryl–reactive blocking agent, a reducing agent, a labeling agent and a detection monoclonal antibody. Each kit has enough reagents to label 40 samples of 100μg each. Similar to the traditional S–nitrosylation switch assay, unmodified cysteines are first blocked using a sulfhydryl–reactive compound (MMTS). S–nitrosylated cysteines are then selectively reduced with ascorbate in HENS Buffer for specific labeling with iodoTMTzero™ Reagents, which irreversibly bind to the cysteine thiol that was S–nitrosylated. Detection of the TMT™ reagent–modified proteins is facilitated using an anti–TMT antibody.
Highlights:
  • Specific – optimized labeling reactions switch S–nitrosocysteine post translational modifications with iodoTMT Reagent for easy detection by Western blot using anti–TMT antibody
  • Better signal-to-noise – uses a non–biological iodoTMT Reagent for labeling instead of HPDP–biotin, resulting in less background during Western blot detection
  • Workflow-compatible – labeling method is compatible with alternative methods of analysis, including enrichment via anti–TMT resin and multiplex quantitation by mass spectrometry
  • Convenient – all needed reagents included one kit
Includes:
Cysteine–blocking reagent, reducing agent, reaction buffer, iodoTMT™ labeling reagent, and anti-TMT antibody