Please login to your online account to display your discounted pricing
Thermo Scientific™ Pierce™ Quantitative Peroxide Assay Kits
Detect and measure hydrogen peroxide levels (H2O2) in biological samples using an iron (Fe) and xylenol orange (XO) reagent for microplates or tubes.
$176.64 - $282.81
Products
Catalog Number Mfr. No. Description Price Quantity    
PI23280
thermo scientific™
PI23280
Pierce Quantitative Peroxide Assay Kit (Aqueous) Pack for $282.81
PI23283
thermo scientific™
23283
Pierce Peroxide Assay Reagent C Each for $176.64
PI23285
thermo scientific™
PI23285
Pierce Quantitative Peroxide Assay Kit (Lipid) Pack for $252.12
Description & Specifications
Detect and measure hydrogen peroxide levels in biological samples using an iron and xylenol orange reagent with Thermo Scientific™ Pierce Quantitative Peroxide Assay Kits.
These aqueous and lipid-compatible peroxide assay kits detect peroxides based on oxidation of ferrous to ferric ion in the presence of xylenol orange. One assay kit has an aqueous-compatible formulation that includes sorbitol, which provides sensitivity enhancement. The other assay kit has a formulation that may be used with lipid-containing samples without first extracting the lipids; however, because this kit does not contain sorbitol, the assay is less sensitive. Most proteins do not interfere with these assays, although some metal chelators may require use of a blank (i.e., control) to mitigate these effects.
In these assays, hydroperoxides convert the Fe2+ to Fe3+ at acidic pH. With the aqueous-compatible formulation, peroxide first reacts with sorbitol, converting it to a peroxyl radical, which in turn initiates Fe2+ oxidation to Fe3+. In the lipid compatible formulation, the peroxide converts the Fe2+ to Fe3+ directly. In a sulfuric acid solution, the Fe3+ complexes with the xylenol orange (XO) dye to yield a purple product having an absorbance maximum at 560nm. Peroxide levels in test samples can be determined by calculation from the know extinction coefficient of the XO-Fe complex or by reference to a standard curve prepared with hydrogen peroxide solution.
Highlights:
  • Peroxides – detects and measures hydrogen peroxide (H2O2) levels in biological and other liquids samples
  • Convenient – no proprietary reagents involved but the ready-to-use kits remove the variability, difficulty and added expense of formulating the reagents yourself
  • Versatile – quantitation by calculation from the extinction coefficient or by reference to a standard curve enables the method to be easily adapted to a variety of sample types and testing purposes
  • Two formulations – choose the aqueous-compatible kit for maximum sensitivity with non-lipid samples; choose the lipid-compatible kit for best results with lipid-containing samples
Recommended for:
General hydrogen peroxide (H2O2) measurement in protein samples; Quantitation of peroxides in detergents; Assessment of the effects of peroxides on cellular activity; Determination of protein glycation; Measurement of peroxide accumulation in lipids