Please login to your online account to display your discounted pricing

Thermo Scientific Pierce™ qIP Protein Interaction Assay Reagents


Prepare optimized buffers for pull-down and reagent sets of TurboLuc (Tluc) assay buffer and coelenterazine substrate for luciferase quantitation with these reagents.

Manufacturer: thermo scientific  82016

 View more versions of this product

Catalog No. PI82016

  • / Each
 

Description & Specifications

Specifications

Quantity 1500-rxn kit
Formulation Two-component kit
Kit Contents Coelenterazine Substrate (100X), 2 x 0.5mL
Sufficient For 1500 luciferase assay reactions

Prepare optimized buffers for pull-down and reagent sets of TurboLuc (Tluc) assay buffer and coelenterazine substrate for luciferase quantitation with Thermo Scientific™ Pierce™ qIP Protein Interaction Reagents.