Please login to your online account to display your discounted pricing

Thermo Scientific Pierce™ qIP Protein Interaction Assay Reagents


Prepare optimized buffers for pull-down and reagent sets of TurboLuc (Tluc) assay buffer and coelenterazine substrate for luciferase quantitation with these reagents.

Manufacturer: thermo scientific  82014

 View more versions of this product

Catalog No. PI82014

  • / Each
 

Description & Specifications

Specifications

Formulation Proprietary aqueous buffer
Quantity 50mL
Sufficient For 500mL 1X Buffer D to dilute lysates and prepare wash buffer for qIP assays

Prepare optimized buffers for pull-down and reagent sets of TurboLuc (Tluc) assay buffer and coelenterazine substrate for luciferase quantitation with Thermo Scientific™ Pierce™ qIP Protein Interaction Reagents.