Please login to your online account to display your discounted pricing

Thermo Scientific Pierce™ qIP Protein Interaction Assay Reagents


Prepare optimized buffers for pull-down and reagent sets of TurboLuc (Tluc) assay buffer and coelenterazine substrate for luciferase quantitation with these reagents.

Manufacturer: thermo scientific  82013

 View more versions of this product

Catalog No. PI82013

  • / Each
 

Description & Specifications

Specifications

Sufficient For 80mL lysis buffer (neat) and 240mL wash buffer (1:3 mixture with 1X Buffer D) for qIP assays
Formulation Proprietary aqueous buffer with detergent
Quantity 140mL

Prepare optimized buffers for pull-down and reagent sets of TurboLuc (Tluc) assay buffer and coelenterazine substrate for luciferase quantitation with Thermo Scientific™ Pierce™ qIP Protein Interaction Reagents.