Please login to your online account to display your discounted pricing
Thermo Scientific™ Pierce™ PVDF Membranes
Enable high protein and nucleic acid binding capacity with sheets and rolls of high-quality polyvinylidene difluoride (PVDF) membrane.
$177.12 - $571.70
Products
Catalog Number Mfr. No. Description Price Quantity    
PI22860
thermo scientific™
22860
Pierce Low-Fluorescence PVDF Transfer Membrane, 0.2um, 7cm x 8.4cm Each for $177.12
PI88518
thermo scientific™
88518
Pierce PVDF Transfer Membrane, 0.45um, 26.5cm x 3.75m Each for $571.70
PI88585
thermo scientific™
88585
Pierce PVDF Transfer Membrane, 0.45um, 10cm x 10cm Each for $234.41
Description & Specifications
Thermo Scientific™ Pierce PVDF Membranes are sheets and rolls of high-quality polyvinylidene difluoride membrane that provide high binding capacity for proteins and nucleic acids for Western, Southern and Northern blotting methods.
These PVDF (polyvinyl difluoride) membranes are more resistant to discoloration than other commercially available PVDF membranes and have better retention of adsorbed proteins than other types of transfer membranes used for Western blotting, including nitrocellulose. The standard 0.45-micron pore size membrane is available as pre-cut sheets and a roll. The Low-Fluorescence PVDF membrane has a 0.2-micron pore size and is ideal for use with fluorescent probes. This membrane has lower background levels and increased sensitivity for fluorescent probing than regular PVDF or nitrocellulose membranes.
Highlights:
  • Western blotting – PVDF transfer membranes manufactured especially for protein transfer and Western blot applications
  • Compatible and durable – polyvinyl difluoride (PVDF) is compatible with most organic solvents, acids and mild bases; does not tear or become brittle like nitrocellulose
  • Easy to use – pre-cut PVDF sheets are ready to use for transfer and blotting experiment from standard precast and homemade mini gels (SDS-PAGE)
  • Specialized options – choose the 0.45μm variety for colorimetric and chemiluminescent detection; use the low-fluorescence membrane (0.2μm) for fluorescent blotting