Please login to your online account to display your discounted pricing
Thermo Scientific™ Pierce™ Protein-Free Blocking Buffers
Block membranes, plates and other samples with these ready-to-use PBS and TBS formulations of a non-protein compound to yield extremely low background.
$63.85 - $311.18
Specifications
Applications ELISA, IHC, WB
View More Specs
Products
Catalog Number Mfr. No. Description Quantity Price Quantity    
PI37570
thermo scientific™
37570
Protein-Free (TBS) Blocking Buffer 1L Each for $288.20
PI37572
thermo scientific™
37572
Protein-Free (PBS) Blocking Buffer 1L Each for $288.20
PI37573
thermo scientific™
37573
Protein-Free T20 (PBS) Blocking Buffer 1L Each for $311.18
PI37584
thermo scientific™
37584
Protein-Free (PBS) Blocking Buffer 100mL Each for $65.48
PI37585
thermo scientific™
37585
Protein-Free (TBS) Blocking Buffer 100mL Each for $63.85
Description & Specifications
Specifications
Applications ELISA, IHC, WB
Thermo Scientific™ Pierce Protein-Free Blocking Buffers are PBS and TBS formulations of a non-protein compound that provides effective blocking for membrane-based and plate-based protein detection methods, resulting in extremely low background.
Traditional blocking buffers contain proteins that can cross-react with immunodetection systems, resulting in high background and reduced signal. Typical commercial alternatives to protein-based blocking agents are ineffective. Pierce Protein-Free Blocking Buffers are devoid of protein while remaining highly effective at blocking plates, membranes and other surfaces for ELISA, Western blotting, glass slide arrays and other applications.
Highlights:
  • Protein-free blocker – minimizes or eliminates cross-reactivity associated with protein-based blocking buffers
  • Application-compatible – effective in all kinds of protein detection systems, including Western blots (membranes), ELISA (microplates) and arrays (coated glass slides)
  • Streptavidin-friendly – absolutely free of biotin; no possible interference with avidin-biotin detection systems
  • High-performance – optimized and validated in many protein methods to provide high signal-to-noise ratio (i.e., no quenching of specific binding and signal but eliminating nonspecific binding and background)
  • Convenient – supplied as ready-to-use 1X formulations in TBS and PBS with and without 0.05% Tween™-20