Thermo Scientific™ Pierce™ Protein Concentrator PES, 2 to 6mL
Thermo Scientific™ Pierce™ Protein Concentrators PES are easy-to-use centrifugal devices that provide fast processing and excellent recovery of protein samples.
$140.00 - $307.31
Specifications
Volume (Metric) 2 to 6mL
View More Specs
Products 12
Catalog Number Mfr. No. MWCO Type Volume (Metric) Quantity Price Quantity  
Catalog Number Mfr. No. MWCO Type Volume (Metric) Quantity Price Quantity  
PI88516 View Documents Thermo Scientific™
88516
10kDa Protein Concentrator 2 to 6mL 10 rxn(s) Each for $152.00
PI88515 View Documents Thermo Scientific™
88515
3kDa Protein Concentrator 2 to 6mL 24 rxn(s) Each for $301.00
PI88514S View Documents Thermo Scientific™
88514S
3K Spin Column, 15 mL conical 2 to 6mL 2pk Each for $140.00
PI88523 View Documents Thermo Scientific™
88523
100K Spin Column, 15 mL conical 2 to 6mL 10pk Each for $152.00
PI88524 View Documents Thermo Scientific™
88524
100K Spin Column, 15 mL conical 2 to 6mL 24pk Each for $301.00
PI88516S View Documents Thermo Scientific™
88516S
10K Spin Column, 15 mL conical 2 to 6mL 2pk Each for $140.00
PI88522 View Documents Thermo Scientific™
88522*
30kDa Protein Concentrator 2 to 6mL 24 rxn(s) Each for $301.00
PI88521 View Documents Thermo Scientific™
88521
30kDa Protein Concentrator 2 to 6mL 10 rxn(s) Each for $152.00
PI88521S View Documents Thermo Scientific™
88521S
30K Spin Column, 15 mL conical 2 to 6mL 2pk Each for $140.00
PI88514 View Documents Thermo Scientific™
88514
3kDa Protein Concentrator 2 to 6mL 10 rxn(s) Each for $152.00
PI88517 View Documents Thermo Scientific™
88517*
10kDa Protein Concentrator 2 to 6mL 24 rxn(s) Each for $307.31
PI88523S View Documents Thermo Scientific™
88523S
100K Spin Column, 15 mL conical 2 to 6mL 2pk Each for $140.00
Description
Description
The Thermo Scientific™ Pierce™ concentrators are easy-to-use centrifugal devices that provide fast processing and excellent recovery of protein samples. These disposable ultrafiltration devices contain a polyethersulfone (PES) membrane for the concentration, desalting, and buffer exchange of biological samples, such as tissue culture media, antiserum, monoclonal antibody preparations, and chromatography fractions. They can also be used to remove unincorporated label following protein modification reactions. 
The PES membrane in these devices is available in five distinct molecular-weight cutoffs (MWCO) of 3K, 5K, 10K, 30K, and 100K, and can be used for processing sample volumes from 100 μL to 100 mL. The MWCOs are etched on the sides of the concentrators for easy identification, while a clear window with graduations on the side of each device allows for visual determination of the concentrated sample (retentate) volume. The unique design provides reliable and consistent results. Multiple device sizes are available to handle maximum sample volumes of 0.5 mL, 6 mL, 20 mL, and 100 mL.
  • Rapid processing—unique design minimizes membrane fouling and sample concentration of 10- to 30-fold can be achieved in 5–30 minutes for 10K MWCO (device dependent—times may vary for other MWCO’s), even with particle laden solutions.
  • High recovery—retain >90% of protein samples while removing contaminants or exchanging buffers.
  • Convenient—clear markings, wide sample chamber and removable filtrate chamber make handling simple and easy.
  • Instrument compatible—can be used with standard centrifuges utilizing either fixed-angle or swinging-bucket rotors.
Specifications
Specifications
2 to 6mL