missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific™ Pierce™ Protein Concentrator PES, 10K MWCO, 20-100 mL

Thermo Scientific™ Pierce™ Protein Concentrators PES are easy-to-use centrifugal devices that provide fast processing and excellent recovery of protein samples.

Manufacturer:  Thermo Scientific™ 88535

Catalog No. PI88535



Description

Description

The Thermo Scientific™ Pierce™ concentrators are easy-to-use centrifugal devices that provide fast processing and excellent recovery of protein samples. These disposable ultrafiltration devices contain a polyethersulfone (PES) membrane for the concentration, desalting, and buffer exchange of biological samples, such as tissue culture media, antiserum, monoclonal antibody preparations, and chromatography fractions. They can also be used to remove unincorporated label following protein modification reactions. 

The PES membrane in these devices is available in five distinct molecular-weight cutoffs (MWCO) of 3K, 5K, 10K, 30K, and 100K, and can be used for processing sample volumes from 100 μL to 100 mL. The MWCOs are etched on the sides of the concentrators for easy identification, while a clear window with graduations on the side of each device allows for visual determination of the concentrated sample (retentate) volume. The unique design provides reliable and consistent results. Multiple device sizes are available to handle maximum sample volumes of 0.5 mL, 6 mL, 20 mL, and 100 mL.
  • Rapid processing—unique design minimizes membrane fouling and sample concentration of 10- to 30-fold can be achieved in 5–30 minutes for 10K MWCO (device dependent—times may vary for other MWCO’s), even with particle laden solutions.
  • High recovery—retain >90% of protein samples while removing contaminants or exchanging buffers.
  • Convenient—clear markings, wide sample chamber and removable filtrate chamber make handling simple and easy.
  • Instrument compatible—can be used with standard centrifuges utilizing either fixed-angle or swinging-bucket rotors.
Specifications

Specifications

Store at room temperature.
Pierce
Room Temperature
Filtration, Polyethersulfone Membrane
Polyethersulfone
4 rxn(s)
Bottle(s), Spin Column
Protein
Yes
20 to 100mL
10kDa
SDS
Documents