Thermo Scientific™ Pierce™ Proteases for Mass Spectrometry
Improve sequence coverage and protein identification for mass spectrometry using these individual, highly specific proteases in custom combinations.
$307.50 - $411.80
Specifications
Formulation Lyophilized enzyme powder
MS-Compatible Yes
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. Description Price Quantity  
Catalog Number Mfr. No. Description Price Quantity  
PI90051 Thermo Scientific™
90051**
Pierce Lys-C Endoproteinase, MS Grade Each for $411.80
PI90053 Thermo Scientific™
90053
Pierce Asp-N Endoproteinase, MS Grade, Each for $351.53
PI90054 Thermo Scientific™
90054*
Pierce Glu-C Endoproteinase, MS Grade Each for $307.50
PI90056 Thermo Scientific™
90056
Pierce Chymotrypsin Endoproteinase, TLCK treated, MS Grade Each for $335.44
Specifications
Description
These Thermo Scientific™ Pierce Proteases are small, convenient packages of individual, purified highly-specific proteases for use in various combinations to obtain better sequence coverage and protein identification for mass spectrometry.
Improvement in sequence coverage and overall protein characterization can result from improving peptide chromatographic or ionization properties. The masses of these larger peptides can be accurately measured at high charge states on popular high-resolution mass spectrometers, reducing the database search time and increasing the sequence coverage and confidence. Finally, larger peptides can be fragmented effectively by new fragmentation techniques like electron transfer dissociation (ETD) or electron capture dissociation (ECD) that leave post-translational modifications, like phosphorylation, intact.
Highlights:
  • Proteases offer specific cleavage at multiple sites, enabling exploration of primary protein structure
  • Better protein characterization results from overlapping peptides with complementary chromatographic, ionization and fragmentation properties
  • Protocols are designed for MS applications
  • Streamlined data processing and analysis improves interpretation and confidence
Description & Specifications
Specifications
Lyophilized enzyme powder
Yes