Please login to your online account to display your discounted pricing

Thermo Scientific™ Pierce™ Protease and Phosphatase Inhibitor Tablets

Ensure complete protection from dephosphorylation and proteolytic degradation with these broad-spectrum protease and phosphatase inhibitor tablets.

$303.45 - $309.75

Specifications

Quantity 20 tablets
Format Tablet
Formulation Inhibitors in proprietary excipient in amber glass vials
Sufficient For 200mL of sample (10mL each mini tablet)
View More Specs
Products
Catalog Number Mfr. No. Description Price Quantity    

PI88668

 
thermo scientific™
88668
Pierce Protease and Phosphatase Inhibitor Mini Tablets Each for $309.75

PI88669

 
thermo scientific™
88669
Pierce Protease and Phosphatase Inhibitor Mini Tablets, EDTA Free Each for $303.45
Description & Specifications

Specifications

Quantity 20 tablets
Format Tablet
Formulation Inhibitors in proprietary excipient in amber glass vials
Sufficient For 200mL of sample (10mL each mini tablet)

Thermo Scientific™ Pierce Protease and Phosphatase Inhibitor Tablets contain both protease inhibitors and phosphatase inhibitors to provide broad-spectrum complete protection from dephosphorylation and proteolytic degradation.

The Pierce Protease and Phosphatase Inhibitor Tablets contain aprotinin, bestatin, E-64 and leupeptin to provide protection from degradation by serine-proteases, cysteine-proteases, aspartic acid-proteases. To prevent the dephosphorylation activity of serine/threonine and tyrosine phosphatases, each tablet also contains sodium fluoride, sodium orthovanadate, sodium pyrophosphate and beta-glycerophosphate. The formulation is available with or without EDTA, a metalloproteinase inhibitor that may interfere with certain downstream applications. Pierce Protease and Phosphatase Inhibitor Tablets are stable at 4°C for up to 12 months, and each tablet is sufficient for 10mL of solution.

Highlights:

  • Complete All-in-one tablet contains both protease and phosphatase inhibitors

  • Easy-to-use the refrigerator-stable tablets are easily dissolved in buffers and lysis reagents

  • Two formulations available with or without EDTA

  • Compatible use with homebrew or commercially available cell lysis reagents and Thermo Scientific Protein Assays

Recommended for:

Protect intact, active cellular proteins from degradation and/or dephosphorylation by endogenous proteases and phosphatases; Preserve protein integrity and phosphorylation upstream of phosphoproteomic analyses and phospho-specific immunoassays