Please login to your online account to display your discounted pricing

Thermo Scientific™ Pierce™ Protease and Phosphatase Inhibitor Tablets


Ensure complete protection from dephosphorylation and proteolytic degradation with these broad-spectrum protease and phosphatase inhibitor tablets.

Manufacturer: thermo scientific™  88668*

 View more versions of this product

Catalog No. PI88668

  • / Each
 

Description & Specifications

Specifications

Format Tablet
Quantity 20 tablets
Description Pierce Protease and Phosphatase Inhibitor Mini Tablets
Composition Sodium fluoride, Sodium orthovanadate, Sodium pyrophosphate, beta-Glycerophosphate, Aprotinin, Bestatin, E-64, Leupeptin, EDTA
Sufficient For 200mL of sample (10mL each mini tablet)
Formulation Inhibitors in proprietary excipient in amber glass vials

Thermo Scientific™ Pierce Protease and Phosphatase Inhibitor Tablets contain both protease inhibitors and phosphatase inhibitors to provide broad-spectrum complete protection from dephosphorylation and proteolytic degradation.

The Pierce Protease and Phosphatase Inhibitor Tablets contain aprotinin, bestatin, E-64 and leupeptin to provide protection from degradation by serine-proteases, cysteine-proteases, aspartic acid-proteases. To prevent the dephosphorylation activity of serine/threonine and tyrosine phosphatases, each tablet also contains sodium fluoride, sodium orthovanadate, sodium pyrophosphate and beta-glycerophosphate. The formulation is available with or without EDTA, a metalloproteinase inhibitor that may interfere with certain downstream applications. Pierce Protease and Phosphatase Inhibitor Tablets are stable at 4°C for up to 12 months, and each tablet is sufficient for 10mL of solution.

Highlights:

  • Complete All-in-one tablet contains both protease and phosphatase inhibitors

  • Easy-to-use the refrigerator-stable tablets are easily dissolved in buffers and lysis reagents

  • Two formulations available with or without EDTA

  • Compatible use with homebrew or commercially available cell lysis reagents and Thermo Scientific Protein Assays

Recommended for:

Protect intact, active cellular proteins from degradation and/or dephosphorylation by endogenous proteases and phosphatases; Preserve protein integrity and phosphorylation upstream of phosphoproteomic analyses and phospho-specific immunoassays