Please login to your online account to display your discounted pricing
Thermo Scientific Pierce™ Power Staining Kits
Get fast Coomassie staining of protein gels with Thermo Scientific™ Pierce™ Power Staining Kits, optimized to work exclusively with the Pierce Power Stain Cassette and Pierce Power Station.
$221.19 - $245.34
Specifications
Kit Contents Destain Solution, 480mL
View More Specs
Products
Catalog Number Mfr. No. Description Formulation Sufficient For Quantity Price Quantity    
PI22839
thermo scientific
22839
Midi Gel Power Staining Kit, 15 Gels, Coomassie Dye, Protein Gel Electrophoresis Application, Colorimetric Detection Method, In-Gel Detection, The Kit Contains Sufficient Reagents To Stain 15 Midi-Sized Gels at 30mL Each Proprietary gel stain and destain solutions 15 Midi-sized gels at 30mL each 15 Gel(s) Each for $221.19
PI22840
thermo scientific
22840
Mini Gel Power Staining Kit, 30 Gels, Coomassie Dye, Protein Gel Electrophoresis Application, Colorimetric Detection Method, In-Gel Detection, The Kit Contains Sufficient Reagents To Stain 30 Mini-Sized Gels at 15mL Each Proprietary gel stain and destain solutions 30 mini-sized gels at 15mL each 30 Gel(s) Each for $245.34
Description & Specifications
Specifications
Kit Contents Destain Solution, 480mL
Get fast Coomassie staining of protein gels with Thermo Scientific™ Pierce™ Power Staining Kits, optimized to work exclusively with the Pierce Power Stain Cassette and Pierce Power Station.
The Pierce Power Staining Kits enable the rapid Coomassie staining of proteins in up to 2 mini polyacrylamide gels and the removal of unbound stain from the gel matrix. Traditional Coomassie staining techniques require staining for one hour to overnight and prolonged destaining for desired results. When used in conjunction with Thermo Scientific Pierce Power Stain Solution, Destain Solution and Gel Pads, the Pierce Power Stainer provides efficient staining and destaining in 6 to 11 minutes with results equivalent to, or better than, traditional Coomassie staining techniques.
Highlights:
  • High performance – performance is equivalent to, or better than, traditional Coomassie staining techniques
  • Fast – optimized ionic solutions enable Coomassie staining of protein and gel destaining within 6 to 11 minutes when used with the Pierce Power Stainer
  • Compatible – verified to work with commonly used precast and homemade SDS-PAGE gels
Includes:
  • Pierce Power Stain Solution, Destain Solution and Gel Pads
Applications:
  • Coomassie staining and destaining for protein visualization in polyacrylamide gels (requires Pierce Power Stain Cassette and Pierce Power Station)
Requires:
  • Pierce Power Stain Cassette and Pierce Power Station or Pierce G2 Fast Blotter (upgraded) with Power Stain Cassette