Thermo Scientific™ Pierce™ PNPP Substrates
Powders, tablets, solutions and complete kits of p-nitrophenyl phosphate, a colorimetric, soluble alkaline phosphatase substrate for use in ELISA applications.
$231.26 - $304.75
Products ${productFamilyLength}
Catalog Number Mfr. No. Description Price Quantity  
Catalog Number Mfr. No. Description Price Quantity  
PI34045 Thermo Scientific™
PI34045
Pierce PNPP Powder Pack for $231.26
PI34047 Thermo Scientific™
PI34047
Pierce PNPP Tablets Pack of 105 Each for $237.55
PI37620 Thermo Scientific™
PI37620
Pierce PNPP Substrate Kit Pack for $304.75
Specifications
Description
Thermo Scientific™ Pierce PNPP Substrates are powders, tablets, solutions and complete kits of p-nitrophenyl phosphate, a colorimetric, soluble substrate of alkaline phosphatase for use in ELISA applications.
PNPP (p-nitrophenyl phosphate, disodium salt) is a widely used substrate for detecting alkaline phosphatase in ELISA applications. When alkaline phosphatase and PNPP are reacted, a yellow water-soluble reaction product is formed. This reaction product absorbs light at 405nm. Pierce PNPP is available in four formats: dry crystalline powder, single-use tablets, a stable substrate solution and a complete kit containing substrate solution and diethanolamine buffer.
Highlights:
  • Absorbance maximum of 505nm for measurement by ELISA plate readers
  • Develops a yellow hue in solution upon reaction with alkaline phosphatase (AP)
  • Safer to use – solution contains a non-mercury, non-azide preservative