Please login to your online account to display your discounted pricing
Thermo Scientific™ Pierce™ Phosphoprotein Enrichment Kit
Purify (enrich) phosphorylated proteins from mammalian cell and tissue lysates using phospho-specific IMAC affinity resin cartidges or kit.
$891.42 - $891.42
Specifications
Support Type Agarose
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    
PI90003
thermo scientific™
90003**
10 column kit Each for $891.42
Description & Specifications
Specifications
Support Type Agarose
Perform efficient enrichment of phosphorylated proteins derived from mammalian cells and tissues with Thermo Scientific™ Pierce Phosphoprotein Enrichment Kit.
The kit is an effective system for isolating the phosphoprotein fraction of mammalian cell and tissue lysates. The included lysis-binding buffer and proprietary affinity resin produce superior yields with negligible non-specific binding. The kit uses a convenient spin column format, and the resulting phosphorylated protein sample is compatible with downstream applications, including Western blotting, mass spectrometry (MS), 2D-PAGE, and protein arrays. The affinity resin is also available as pre-packed chromatography cartridges for use with automated liquid chromatography (LC) systems.
Highlights:
  • Specific – low nonspecific protein contamination from complex biological samples, such as cell culture lysate and mouse tissue extract
  • Fast – easy spin format enables enrichment of phosphorylated proteins in less than 2 hours
  • Superior yield – achieves higher yields than other commercially available phosphoprotein isolation kits
  • Convenient format – complete kit offering pre-dispensed spin columns, buffers, reagents and ultrafiltration columns for enrichment
  • Compatible – downstream applications include mass spectrometry, Western blotting and 2D-PAGE