missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Thermo Scientific™ OPD (o-phenylenediamine dihydrochloride)
Water-soluble horseradish peroxidase (HRP) substrate that produces a yellow-orange product detectable at 492nm by ELISA plate readers.
Supplier: Thermo Scientific™ 34005
Description
Thermo Scientific™ Pierce OPD is a water-soluble substrate for horseradish peroxidase (HRP) that produces a yellow-orange product detectable at 492nm by ELISA plate readers.
OPD (o-phenylenediamine dihydrochloride) yields a water-soluble yellow-orange product when reacted with peroxidase with an absorbance maximum of 492nm. OPD can easily be dissolved in a substrate buffer such as Thermo Scientific Stable Peroxide Substrate Buffer (Part No. 34062).Highlights:
- Water soluble product when reacted with peroxidase
- Absorbance maximum of 492nm
Specifications
Pierce OPD Powder | |
Colorimetric | |
HRP (Horseradish Peroxidase) |
ELISA Substrate | |
Chemical Substrate | |
Upon receipt store product at -20°C. Product is shipped at ambient temperature. |
For Research Use Only. Not for use in diagnostic procedures.