Thermo Scientific™ Pierce™ OPD Substrates
Water-soluble horseradish peroxidase (HRP) substrate that produces a yellow-orange product detectable at 492nm by ELISA plate readers.
$116.69 - $243.85
Products ${productFamilyLength}
Catalog Number Mfr. No. Description Price Quantity  
Catalog Number Mfr. No. Description Price Quantity  
PI34005 Thermo Scientific™
PI34005
Pierce OPD Powder Pack for $116.69
PI34006 Thermo Scientific™
PI34006
Pierce OPD Tablets Pack of 50 Each for $243.85
Specifications
Description
Thermo Scientific™ Pierce OPD is a water-soluble substrate for horseradish peroxidase (HRP) that produces a yellow-orange product detectable at 492nm by ELISA plate readers.
OPD (o-phenylenediamine dihydrochloride) yields a water-soluble yellow-orange product when reacted with peroxidase with an absorbance maximum of 492nm. OPD can easily be dissolved in a substrate buffer such as Thermo Scientific Stable Peroxide Substrate Buffer (Part No. 34062).
Highlights:
  • Water soluble product when reacted with peroxidase
  • Absorbance maximum of 492nm