Please login to your online account to display your discounted pricing

Thermo Scientific™ Normal Serum

Normal Serum Control from Thermo Scientific™ Pierce™ is validated for BLOCK, IF and IHC.

$55.10 - $55.10

Specifications

Applications IHC
Formulation 120mg unpurified control antibody in whole serum diluted in PBS and 0.05% sodium azide; pH 7.6
View More Specs
Products
Catalog Number Mfr. No. Description Quantity Price Quantity    

PI31890

 
pierce biotechnology inc
31890
Normal Serum, Swine 120mg Each for $55.10
Description & Specifications

Specifications

Applications IHC
Formulation 120mg unpurified control antibody in whole serum diluted in PBS and 0.05% sodium azide; pH 7.6

Normal Serum Control from Thermo Scientific™ Pierce™ is validated for BLOCK, IF and IHC.

Normal Sera are non-immune serum samples from goat, human, mouse, rabbit and other species that contain the usual complement of serum proteins, including the various classes of immunoglobulins that exist in healthy animals of the particular species. Normal serum provides a sufficient quantity of endogenous proteins to saturate and block nonspecific binding sites on fixed samples or assay substrates, making it useful for immunodetection methods, especially those involving tissue samples such as immunohistochemistry (IHC). Normal sera are also useful as controls for testing general and specific antibody purification methods. When used as an antibody diluent, non-immune serum also supplies a native chemical environment for antibodies to function properly without adding foreign protein components.

Formulation: 120mg of Unpurified control in whole serum, with PBS diluted in PBS; pH 7.6.
31874 has been successfully used in BLOCK, IF and IHC procedures.