Thermo Scientific™ Pierce™ Nitrocellulose Membranes
Sheets and rolls of nitrocellulose, available in two pore sizes, with excellent binding properties for Western blot and other protein or nucleic acid methods.
$184.45 - $587.32
Specifications
Formulation 100% nitrocellulose paper
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. Description Price Quantity  
Catalog Number Mfr. No. Description Price Quantity  
PI77010 Thermo Scientific™
PI77010
Pierce Nitrocellulose Membrane, 0.45um, 8cm x 12cm Pack of 25 Each for $443.94
PI77012 Thermo Scientific™
PI77012
Pierce Nitrocellulose Membrane, 0.2um, 8cm x 12cm Pack of 25 Each for $447.35
PI88013 Thermo Scientific™
PI88013
Pierce Nitrocellulose Membrane, 0.2um, 7.9cm x 10.5cm Pack of 15 Each for $188.92
PI88018 Thermo Scientific™
PI88018
Pierce Nitrocellulose Membrane, 0.45um, 30cm x 3.5m Pack for $587.32
PI88014 Thermo Scientific™
PI88014
Pierce Nitrocellulose Membrane, 0.45um, 7.9cm x 10.5cm Pack of 15 Each for $203.37
PI88024 Thermo Scientific™
PI88024
Pierce Nitrocellulose Membrane, 0.2um, 8cm x 8cm Pack of 15 Each for $184.45
PI88025 Thermo Scientific™
PI88025
Pierce Nitrocellulose Membrane, 0.45um, 8cm x 8cm Pack of 15 Each for $184.45
Specifications
Description
Thermo Scientific™ Pierce Nitrocellulose Membranes are sheets and rolls of nitrocellulose, available in two pore sizes, that have excellent binding properties for Western blotting, dot-blot assays and other protein or nucleic acid methods.
Nitrocellulose is a popular binding matrix for Western blotting because of its high affinity for proteins and compatibility with a variety of detection methods (e.g., chemiluminescence, chromogenic, fluorescence). Pierce Nitrocellulose Membranes with 0.2 and 0.45 micron pore sizes are available, as are a variety of package sizes containing pre-cut sheets for nearly any application.
Highlights:
  • High-quality – pure, 100% nitrocellulose membranes with high surface area and excellent uniformity
  • Choices – available in 0.2μm and 0.45μm pore sizes, nitrocellulose varieties for peptide- and protein-applications, respectively
  • Convenient – several sizes of pre-cut sheets and an economically-priced roll for cutting to any dimensions
  • High-binding affinity – provides excellent protein binding, blocks easily, and produces very low background in chemiluminescent Western blotting
Description & Specifications
Specifications
100% nitrocellulose paper