Thermo Scientific™ Pierce™ Nickel Coated Plates
Bind His-tagged fusion proteins for ELISA and other plate-based assays to study interactions of expressed recombinant proteins.
$339.48 - $362.04
Specifications
Binding Target His tag
Quantity 5 plates
Sufficient For Binding approx. 9pmol His-tagged protein per well
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. Description Price Quantity  
Catalog Number Mfr. No. Description Price Quantity  
PI15142 Thermo Scientific™
PI15142
Pierce Nickel Coated Plates, Clear, 96-Well Strip Pack of 5 Each for $339.48
PI15242 Thermo Scientific™
PI15242
Pierce Nickel Coated Plates, White, 96-Well Pack of 5 Each for $361.57
PI15342 Thermo Scientific™
PI15342
Pierce Nickel Coated Plates, Black, 96-Well Pack of 5 Each for $362.04
PI15442 Thermo Scientific™
15442*
Pierce Nickel Coated Plates, Clear, 96-Well Pack of 5 Each for $350.62
Specifications
Description
Thermo Scientific™ Pierce Nickel Coated Plates provide a simple format to bind His-tagged fusion proteins for ELISA and other plate-based assays involving protein interactions of expressed recombinant proteins.
These Ni(2+) chelate-coated plates are ideal for analyzing polyhistidine-tagged fusion proteins by ELISA-based methods. Proteins that contain a succession of several histidine residues at the amino or carboxyl terminus have a strong binding affinity for metal. Bacterial lysates containing polyhistidine-tagged fusion proteins can be added directly to the plates without the need for blocking. The clear, white or black plates can be used with colorimetric, chemiluminescent or fluorescent detection methods, respectively.
Highlights:
  • Detergents used to lyse cells do not inhibit binding to Ni2+coated plates as they do with plain polystyrene
  • The detection limit is 1ng of histidine fusion protein
  • Better binding for more sensitive assays compared to other commercially available nickel-activated plates
Description & Specifications
Specifications
His tag
5 plates
Binding approx. 9pmol His-tagged protein per well