Thermo Scientific™ Pierce™ NHS-Phosphine Staudinger Ligation Reagents
Derivatize proteins or amine surfaces for selective conjugation, detection or purification of azide-tagged targets with these activated phosphine compounds.
$535.19 - $535.19
Specifications
Chemical Reactivity Amine-to-Nonselective
Cleavable No
Quantity 10mg
PEGylated No
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. Description Price Quantity  
Catalog Number Mfr. No. Description Price Quantity  
PI88900 Thermo Scientific™
88900
Pierce NHS-Phosphine Each for $535.19
Specifications
Description
Thermo Scientific™ Pierce NHS-Phosphine and Sulfo-NHS-Phosphine are amine-reactive compounds that can be used to derivatize primary amines of proteins or amine-coated polymer surfaces for ligation to azide-tagged molecules.
These NHS-ester compounds react to form covalent bonds with primary amines (e.g., side chain of lysine residues or aminosilane-coated surfaces). The phosphine groups (PPh2) react with azide-labeled molecules by a mechanism known as Staudinger chemistry, enabling efficient and specific conjugation of derivatized molecules in biological samples. In combination with different azide-tagged compounds, these NHS-Phosphine reagents facilitate a variety of labeling and crosslinking experimental strategies for investigating protein interactions and cellular pathways.
Highlights:
  • Versatile – Sulfo-NHS-Phosphine easily dissolves in water-miscible solvents for subsequent dilution in aqueous reaction mixtures for azide conjugation; NHS-Phosphine can be used for amine surface modification in organic solvents
  • Compatible – reaction chemistry occurs effectively in simple buffer conditions; requires no accessory reagents such as copper or reducing agents
  • Specific – NHS esters are specific for covalent attachment to primary amines (side chain of lysine or N-terminus of polypeptides)
  • Chemoselective – azide and phosphine groups do not react or interfere with components of biological samples but conjugate to one another with high efficiency
Description & Specifications
Specifications
Amine-to-Nonselective
No
10mg
No