Thermo Scientific™ Pierce™ NHS-Diazirine Crosslinkers
Very short to mid-length (3.9 to 13.5A), amine-to-anything crosslinkers; NHS-ester and diazirine reactive groups; photoactivate with UV light (Amax 330 to 370nm).
$363.95 - $552.35
Specifications
Chemical Reactivity Amine-to-Nonselective
Quantity 50mg
PEGylated No
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. Description Price Quantity  
Catalog Number Mfr. No. Description Price Quantity  
PI26167 Thermo Scientific™
26167
Pierce SDA Each for $363.95
PI26168 Thermo Scientific™
26168
Pierce LC-SDA Each for $495.90
PI26169 Thermo Scientific™
26169
Pierce SDAD Each for $552.35
PI26174 Thermo Scientific™
26174
Pierce Sulfo-LC-SDA Each for $490.10
Specifications
Description
Thermo Scientific™ Pierce NHS-Diazirine Crosslinkers combine proven NHS-ester and diazirine-based photoreaction chemistries to conjugate amine-containing molecules with nearly any other functional group via long-wave UV-light activation.
The succinimidyl-ester diazirine (SDA) reagents are a new class of crosslinkers that combine proven amine-reactive chemistry with an innovative and efficient diazirine-based photochemistry for conjugating amine-containing molecules to nearly any other functional group. The SDA crosslinkers include six compounds differing in spacer arm lengths, ability to cleave the crosslinked proteins, and presence or absence of a charged group for membrane permeability. Protein crosslinking is an important technique used to understand protein structure and to stabilize protein-protein interactions. Thermo Scientific SDA Reagents extend the efficiency and range of interactions that can be explored by this approach.
Highlights:
  • Versatile - different amine-reactive diazirine derivatives capture specific intracellular or cell-surface protein interactions
  • Controllable - two-step chemical crosslinking is activated using common laboratory UV lamps
  • Reversible - disulfide-containing spacers enable cleavage using reducing agents
  • Easy - reagents are photo-stable under typical laboratory lighting conditions so there is no need to perform experiments in the dark
  • Better than aryl azides - diazirine photo-reactive group has better photostability and is more efficiently activated by long-wave UV light
Description & Specifications
Specifications
Amine-to-Nonselective
50mg
No