Please login to your online account to display your discounted pricing
Thermo Scientific™ Pierce™ NHS-Azide Staudinger Ligation Reagents
Activated azido compounds to derivatize proteins or amine surfaces for selective conjugation, detection or purification by phosphine-modified probes.
$370.10 - $584.61
Specifications
Chemical Reactivity Amine-to-Nonselective
Cleavable No
Membrane Permeable Yes
Water Soluble No
View More Specs
Products
Catalog Number Mfr. No. Description Price Quantity    
PI26130
thermo scientific™
26130
Pierce NHS-PEG4-Azide Each for $584.61
PI88902
thermo scientific™
88902
Pierce NHS-Azide Each for $370.10
Description & Specifications
Specifications
Chemical Reactivity Amine-to-Nonselective
Cleavable No
Membrane Permeable Yes
Water Soluble No
Thermo Scientific™ Pierce NHS-Azide reagents are amine-reactive compounds that can be used to derivatize primary amines of proteins or amine-coated polymer surfaces for ligation to phosphine-modified molecules.
These NHS-ester compounds react to form covalent bonds with primary amines (e.g., side chain of lysine residues or aminosilane-coated surfaces). The azide (N3) group reacts with phosphine-labeled molecules by a mechanism known as Staudinger chemistry, enabling efficient and specific conjugation of derivatized molecules in biological samples. In combination with different phosphine-activated compounds, these NHS-Azide reagents facilitate a variety of labeling and crosslinking experimental strategies for investigating protein interactions and cellular pathways.
Highlights:
  • Soluble – reagents easily dissolve in water-miscible solvents for subsequent dilution in aqueous reaction mixtures with cell lysates and other biological samples
  • Compatible – reaction chemistry occurs effectively in simple buffer conditions; requires no accessory reagents such as copper or reducing agents
  • Specific – NHS esters are specific for covalent attachment to primary amines (side chain of lysine or N-terminus of polypeptides)
  • Chemoselective – azide and phosphine groups do not react or interfere with components of biological samples but conjugate to one another with high efficiency
  • Pegylated – choose the very short NHS-Azide or one of the two varieties whose spacer arms contain medium (4-unit) or long (12-unit) polyethylene glycol (PEG) groups for increased solubility and reach