missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific™ Pierce™ NBT/BCIP Substrate Solutions

Powders and ready-made nitro-blue tetrazolium and 5-bromo-4-chloro-3'-indolyphosphate solutions for chromogenic detection of alkaline phosphatase probes.

$166.46 - $381.21

Specifications

Detection Chromogenic
Storage Requirements 4°C
Applications Alkaline phosphate detection
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Description Price Quantity  
Catalog Number Mfr. No. Description Price Quantity  
PI34035 Thermo Scientific™
PI34035
Pierce NBT Substrate Pack for $224.73
PI34040 Thermo Scientific™
PI34040
Pierce BCIP Substrate Pack for $381.21
PI34042 Thermo Scientific™
PI34042
Pierce 1-Step NBT/BCIP Substrate Solution Pack for $229.48
PI34070 Thermo Scientific™
PI34070
Pierce 1-Step NBT/BCIP Plus Suppressor Substrate Solution Pack for $166.46
Specifications

Description

Thermo Scientific™ Pierce NBT and BCIP Substrates are powders and ready-made solutions of nitro-blue tetrazolium and 5-bromo-4-chloro-3'-indolyphosphate for chromogenic blot and IHC staining with alkaline phosphatase probes.

The combination of NBT (nitro-blue tetrazolium chloride) and BCIP (5-bromo-4-chloro-3'-indolyphosphate p-toluidine salt) yields an intense, insoluble black-purple precipitate when reacted with alkaline phosphatase, a popular enzyme conjugate for antibody probes. Pierce NBT and BCIP powders are available individually and in two convenient, ready-to-use solutions (with and without a levamisole suppressor of endogenous phosphatase activity).

Highlights:

  • AP substrate – for detection of alkaline phosphatase (alk-phos) activity on solid media, including nitrocellulose and PVDF membranes and fixed tissue samples

  • Chromogenic – no special equipment needed for visualization; produces purple-black precipitate that is easy to photograph

  • Package options – choose individual NBT and BCIP powders, or preformulated, ready-to-use solutions with or without levamisole suppressor

1-Step™ NBT/BCIP

  • Ideal for immunoblotting applications
  • Minimal assay-to-assay variability

1-Step NBT/BCIP Plus Suppressor

  • Ideal for staining tissues assayed with calf intestinal alkaline phosphatase without the interference caused by endogenous phosphatases
  • Contains 1mM levamisole to inhibit endogenous phosphatase activity
Description & Specifications

Specifications

Chromogenic
4°C
Alkaline phosphate detection