Thermo Scientific™ Pierce™ Micro BCA™ Protein Assay
Special 3-component version of our popular BCA Reagents to measure total protein concentration of dilute protein solutions (0.5 to 20 micrograms/mL).
$57.29 - $346.87
Products ${productFamilyLength}
Catalog Number Mfr. No. Description Price Quantity  
Catalog Number Mfr. No. Description Price Quantity  
PI23231 Thermo Scientific™
PI23231
Pierce Micro BCA Reagent A (MA) Pack for $96.46
PI23232 Thermo Scientific™
PI23232
Pierce Micro BCA Reagent B (MB) Pack for $170.22
PI23234 Thermo Scientific™
PI23234
Pierce Micro BCA Reagent C (MC) Pack for $57.29
PI23235 Thermo Scientific™
PI23235
Pierce Micro BCA Protein Assay Kit Pack for $346.87
Specifications
Description
Thermo Scientific™ Pierce Micro BCA Protein Assay uses a concentrated form of the BCA Reagents to accurately measure dilute protein solutions (0.5 to 20μg/mL).
Additional Information and Online Ordering »
The Micro BCA Protein Assay Kit is a specialized version of the popular Pierce BCA Protein Assay for determining the protein concentration of dilute samples. Mixing together the three Micro BCA Reagents results in a working solution that is sufficiently concentrated to measure protein when mixed with an equal volume of sample. The result is an assay for accurately measuring 0.5 to 20μg/mL protein solutions. The assay is exceptionally linear and exhibits very low levels of protein-to-protein variability.
Highlights:
  • Sensitive – accurately detect down to 0.5μg/mL (2μg/mL in microplate format)
  • High linearity – linear working range for BSA equals 0.5 to 20μg/mL
  • Colorimetric – measure with a standard spectrophotometer or plate reader (562nm)
  • Compatible – unaffected by typical concentrations of most ionic and nonionic detergents
  • Convenient – microplate and cuvette protocols provided with the instructions
  • Stable – kit stable at room temperature; prepared working reagent stable for to 24 hours
Includes:
Three Micro BCA Reagents (MA, MB, MC) and 10 ampules of Albumin Standard (2mg/mL)