Please login to your online account to display your discounted pricing
Thermo Scientific™ Pierce™ Methyl-PEG-Maleimide Reagents
Pegylate sulfhydryls (-SH groups) on proteins (cysteine residues) or assay surfaces using MS(PEG); sulfhydryl-reactive (maleimide), PEG compounds (12 and 24 EG units).
$542.99 - $565.50
Specifications
Capping Group Methyl
Chemical Reactivity Sulfhydryl
Cleavable No
PEGylated Yes
View More Specs
Products
Catalog Number Mfr. No. Description Quantity Price Quantity    
PI22711
thermo scientific™
22711
Pierce MM(PEG)12 100mg Each for $542.99
PI22713
thermo scientific™
22713
Pierce MM(PEG)24 100mg Each for $565.50
Description & Specifications
Specifications
Capping Group Methyl
Chemical Reactivity Sulfhydryl
Cleavable No
PEGylated Yes
Thermo Scientific™ Pierce MM(PEG)12 is a methyl-terminated, polyethylene glycol compound (3 times 12 PEG units) activated with a maleimide group for covalent pegylation of sulfhydryls on proteins (e.g., cysteines) or assay surfaces.
MM(PEG)n is the abbreviation for a set of compounds having polyethylene glycol (PEG) spacers with methyl (—CH3) and sulfhydryl-reactive maleimide groups at opposite ends. The unbranched, hydrophilic, discrete-length molecules have the form Methyl-PEGn-Maleimide, where the subscript “n” denotes 12 or 24 ethylene glycol units. The maleimide is spontaneously reactive with sulfhydryl (—SH) groups, providing for efficient PEGylation of proteins, peptides and other cysteine- or thiol-containing molecules.
Highlights:
  • Maleimide-activated for efficient PEGylation of sulfhydryl groups at pH 6.5-7.5; reaction of maleimide group results in formation of stable, irreversible thioether bonds
  • Fully characterized PEGylation reagents with defined PEG chain lengths; molecules of discrete molecular weight for consistency of performance in protein-modification applications
  • Provided with either 12 or 24 ethylene glycol units, enabling modification procedures to be optimized for a specific application while retaining all the benefits associated with protein PEGylation
  • PEG spacer provides unique advantages, including increased stability, reduced tendency toward aggregation and reduced immunogenicity
  • Easy-to-follow instructions increase the likelihood of a successful outcome