Thermo Scientific™ Pierce™ Mass Spec Sample Prep Kit for Cultured Cells
Perform preparation of clean peptide mixtures from cultured cells for mass spectrometry (MS) analysis with this easy-to-use, comprehensive kit.
Manufacturer:  Thermo Scientific™ 84840
 View more versions of this product
Catalog No. PI84840
Specifications
Description
Perform preparation of clean peptide mixtures from cultured cells for mass spectrometry (MS) analysis with Thermo Scientific™ Pierce Mass Spec Sample Prep Kit for Cultured Cells, an easy-to-use, comprehensive kit.
Additional Information and Online Ordering »
The Pierce Mass Spec Sample Prep Kit for Cultured Cells contains all the necessary reagents and enzymes to prepare up to 20 samples (1 million cells each) for MS analysis. The simple and robust workflow uses a lysis protocol that generates approximately 100μg of protein per sample and includes a control digestion indicator to monitor the efficiency of the two-step enzymatic digestion protocol. The kit procedure, which includes a Digestion Indicator as an internal protein/peptide control, produces consistently and reproducibly clean peptide mixtures for protein identification.
Highlights:
  • Complete – includes all reagents, a digestion indicator control, proteases and an optimized protocol needed to process up to 20 samples
  • Simple – user friendly kit can provide reproducible results even for non-expert MS analysts
  • Flexible – can be adapted to handle sample sizes between 10 to 200μg
  • High yield – total protein yield from 1 million cells is greater than 100μg
  • Optimized – cysteine reduction and alkylation are 100% with less than 1% over alkylation of non-cysteine residues
  • Efficient - percentage of missed cleavages is less than 10%
  • Compatible – final preparation is ready for direct MS analysis and other downstream applications, including mass-tag labeling
Includes:
Kit contains proprietary lysis and digestion buffers, digestion indicator, DTT, iodoacetamide, Lys-C, trypsin
Description & Specifications
Specifications
Cell Lysis Buffer, 5mL
20 protease digestions and preparations from 100µg of cell lysate
Multi-component kit
20 rxn kit
Pierce Mass Spec Sample Prep Kit for Cultured Cells
Promotions
Promotions
  • To make a purchase through the 2016 Thermo Scientific™ Mass Spec Rewards Program, select a combination of products from this list within and contact your local sales representative for a quote. Please mention the appropriate Promo Code from the table within.
    12/31/17