Thermo Scientific™ Pierce™ Maleimide Activated (Sulfhydryl-binding) Plates
Covalently attach sulfhydryl-containing molecules that are difficult to coat onto polystyrene plates, such as peptides that contain a terminal cysteine.
$239.06 - $253.76
Specifications
Binding Target Sulfhydryl
Quantity 5 plates
Sufficient For Binding approx. 100 to 150pmol peptide-SH per well
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. Description Price Quantity  
Catalog Number Mfr. No. Description Price Quantity  
PI15150 Thermo Scientific™
PI15150
Pierce Maleimide Activated Plates, Clear, 96-Well Strip Pack of 5 Each for $239.06
PI15153 Thermo Scientific™
15153
Pierce Maleimide Activated Plates, Black, 96-Well Pack of 5 Each for $253.76
PI15152 Thermo Scientific™
15152
Pierce Maleimide Activated Plates, White, 96-Well Pack of 5 Each for $253.76
Specifications
Description
Thermo Scientific™ Pierce Maleimide Activated Plates are ideal for binding sulfhydryl-containing molecules that are difficult to coat onto polystyrene plates, such as peptides that contain a terminal cysteine.
Pierce Maleimide Activated Plates are provide covalent attachment (plate capture) of sulfhydryl-containing molecules, such as cysteine-terminated peptides. These coated plates are an especially useful tool for assessing specific anti-hapten antibody titers during antibody production. Maleimide Activated Plates are available in clear for colorimetric assays, white for chemiluminescent assays and black for fluorescent assays.
Highlights:
  • Spontaneous immobilization of cysteine-containin peptides and proteins with free sulfhydryls
  • Pre-blocked to reduce nonspecific binding
Description & Specifications
Specifications
Sulfhydryl
5 plates
Binding approx. 100 to 150pmol peptide-SH per well