Thermo Scientific™ Pierce™ Maleimide-Activated HRP Conjugation Kit
Activated horseradish peroxidase (HRP) enzyme and kit to label proteins, peptides or other ligands via sulfhydryl groups (reduced cysteines) to make HRP conjugates.
$316.10 - $557.01
Products ${productFamilyLength}
Catalog Number Mfr. No. Formulation Price Quantity  
Catalog Number Mfr. No. Formulation Price Quantity  
PI31485 Thermo Scientific™
PI31485
Activated horseradish peroxidase lyophilized from PBS Pack for $316.10
PI31494 Thermo Scientific™
PI31494
Multi-component kit with activated HRP Pack for $557.01
Specifications
Description
Thermo Scientific™ Pierce Maleimide-Activated HRP Conjugation Kit can be used to directly prepare HRP conjugates from proteins, peptides or other ligands that contain sulfhydryl groups, such as reduced cysteines.
The Thermo Scientific Maleimide Activated Horseradish Peroxidase (HRP) Kit can be used to directly prepare HRP conjugates from proteins, peptides or other ligands that contain a free sulfhydryl group. This kit eliminates the first step of the two-step maleimide method, making conjugate preparation much simpler. The kit is supplied with a stable, Maleimide-activated horseradish peroxidase. HRP conjugates can be purified by gel filtration chromatography, ultrafiltration or dialysis, depending upon the size of the conjugated protein.
Highlights:
  • Prepare enzyme conjugates from proteins that contain a free sulfhydryl
  • Includes 2-MEA to generate free sulfhydryls from disulfide bonds
  • Includes SATA to add free sulfhydryls to lysine residues