Thermo Scientific™ Pierce™ MALDI Matrices
Highly purified, recrystallized CHCA, DHB and SA reagents in single-use tubes for mass spectrometry by matrix-assisted laser desorption ionization (MALDI).
$239.25 - $297.41
Specifications
Quantity 24 x 1mg
Sufficient For Analysis of up to 96 samples with each tube after dissolving its contents in 100µL of solvent
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. Description Price Quantity  
Catalog Number Mfr. No. Description Price Quantity  
PI90031 Thermo Scientific™
90031
Pierce CHCA MALDI Matrix Pack of 24 Each for $270.23
PI90032 Thermo Scientific™
90032
Pierce SA MALDI Matrices Pack of 24 Each for $239.25
PI90033 Thermo Scientific™
90033
Pierce DHB MALDI Matrices Pack of 24 Each for $297.41
PI90035 Thermo Scientific™
90035
Pierce MALDI Matrix Sample Pack Pack of 24 Each for $270.23
Specifications
Description
Thermo Scientific™ MALDI Matrices are highly purified, recrystallized reagents packaged in convenient single-use tubes sufficient for analyzing up to 96 samples by matrix assisted laser desorption ionization.
The MALDI method for MS peptide analysis is now easier to perform with single-use, highly pure MALDI matrices that are exceptionally convenient to use. Matrix assisted laser desorption ionization (MALDI) matrices produce the cleanest mass spectrometric (MS) spectra when they are recrystallized and prepared daily, which is inconvenient and typically results in significant waste. The packaging of purified, recrystallized alpha-cyano-4-hydroxy-cinnamic acid (CHCA), sinapinic acid (SA) and 2,5-dihydroxybenzoic acid (DHB) MALDI matrices makes preparation of high-quality MALDI reagents easy.
Highlights:
  • Highly purified, recrystallized reagent
  • Packaged in convenient single-use tubes sufficient for analyzing up to 96 samples to save time and reduce waste
  • Provides accurate quantity of matrix every time
Description & Specifications
Specifications
24 x 1mg
Analysis of up to 96 samples with each tube after dissolving its contents in 100µL of solvent