Please login to your online account to display your discounted pricing
Thermo Scientific™ Pierce™ Kinase Enrichment Kits and Probes
Desthiobiotin-ATP and -ADP probes to label, affinity purify and measure the active site of ATPases (kinases), including chaperones and metabolic enzymes.
$605.22 - $1,229.46
Specifications
Sufficient For 16 pull-down assays
View More Specs
Products
Catalog Number Mfr. No. Formulation Quantity Price Quantity    
PI88311
thermo scientific™
88311
Desthiobiotin-ATP, MW 1259 16 x 12.6µg Each for $605.22
PI88312
thermo scientific™
88312*
Multi-component kit with Desthiobiotin-ADP Probe 16 rxn kit Each for $1,229.46
PI88313
thermo scientific™
88313
Desthiobiotin-ADP, MW 994 16 x 9.9µg Each for $611.78
PI88310
thermo scientific™
88310
Multi-component kit with Desthiobiotin-ATP Probe 16 rxn kit Each for $923.57
Description & Specifications
Specifications
Sufficient For 16 pull-down assays
Thermo Scientific™ Pierce Kinase Enrichment Kits utilize ActivX™ ATP or ADP Probes to covalently label the active site of ATPases, including chaperones and metabolic enzymes, to enable their selective enrichment using a desthiobiotin tag.
These products, based on ActivX ATP and ADP Probes, feature an amine-reactive nucleotide analog and a desthiobiotin (biotin analog) tag that facilitates selective labeling of lysines in the kinase active site and then subsequent enrichment and recovery of labeled protein. These features allow identification and profiling of target enzyme classes across samples or assessment of the specificity and affinity of enzyme inhibitors.
Highlights:
  • Specific – label only the conserved active-site lysines of nucleotide-binding proteins
  • Flexible – use for in vitro labeling of ATPase enzymes derived from cells or tissues
  • Compatible – use with Western blot or mass spectrometry (MS) workflows
Recommended for:
Profile small-molecule binding affinities and active-site inhibition in a dose-dependent manner; Identify dozens to hundreds of inhibitor targets and off-targets from tissues, cells and subcellular proteomes; Enrichment of enzymes based on function.