Please login to your online account to display your discounted pricing
Thermo Scientific™ Pierce™ Iodoacetic Acid
Get rapid carboxymethylation of reduced cysteine residues with Thermo Scientific™ Pierce™ Iodoacetic Acid and Iodoacetamide.
$137.26 - $254.02
Specifications
Chemical Reactivity Sulfhydryl
View More Specs
Products
Catalog Number Mfr. No. Description Price Quantity    
PI35603
thermo scientific™
PI35603
Iodoacetic Acid Pack for $137.26
PI90034
thermo scientific™
90034
Iodoacetamide, Single-Use Each for $254.02
Description & Specifications
Specifications
Chemical Reactivity Sulfhydryl
Get rapid carboxymethylation of reduced cysteine residues with Thermo Scientific™ Pierce™ Iodoacetic Acid and Iodoacetamide.
Iodoacetic Acid (IAA) can react with several protein functional groups but is typically used for specific S-carboxymethylation of sulfhydryls (reduced cysteines).
Iodoacetic acid reacts with sulfhydryls on cysteines, imidazolyl side chain nitrogens of histidines, the thioether of methionine and the primary amine group of lysines.The rate of reaction and specificity is dependent on the ionization level, which can be manipulated by the pH of the reaction condition.
Highlights:
  • React at slightly alkaline pH for specific S-carboxymethylation of free sulfhydryls
  • React at low pH for specific carboxymethylation of methionines
  • React at high pH to favor carboxymethylation of histidines and lysines
  • Methylate reduced cysteine peptide fragments in protease digests for mass spectrometry