Thermo Scientific™ Pierce™ High Sensitivity Streptavidin Coated Plates
Maximize biotinylated protein capture efficiency and control background with microplates coated to detect low concentrations of biotinylated analytes.
$366.22 - $366.35
Specifications
Binding Target Biotin
Quantity 5 plates
Sufficient For Detection with low-concentration biotin samples
View More Specs
Alert:
Products ${productFamilyLength}
Catalog Number Mfr. No. Description Price Quantity  
Catalog Number Mfr. No. Description Price Quantity  
PI15520 Thermo Scientific™
15520
Pierce Streptavidin Coated High Sensitivity Plates, Clear, 96-Well Strip Each for $366.22
PI15525 Thermo Scientific™
15525
Pierce Streptavidin Coated High Sensitivity Plates, Black, 96-Well Strip Each for $366.35
Specifications
Description
Thermo Scientific™ Pierce High Sensitivity Streptavidin Plates are biotin-binding microplates that are coated to maximize capture efficiency of biotinylated probes and control background in assays for low levels of target analyte.
These high sensitivity plates enable detection of low concentrations (ng/mL) of biotinylated antibodies and other proteins. Whether detecting a signal by fluorescent, colorimetric or chemiluminescent methods, these plates make it easy to develop an ELISA for any sample with a low-level target.
Highlights:
  • Sensitive – detect down to 5ng/mL of biotinylated IgG
  • Specific – low background with high signal-to-noise ratios
  • Versatile – use either fluorescence, chemiluminescence or colorimetric detection
  • Robust – broad dynamic range
  • Ready to use – plates are supplied pre-blocked to save time
Description & Specifications
Specifications
Biotin
5 plates
Detection with low-concentration biotin samples