Thermo Scientific™ Pierce™ High Capacity Copper Coated Plates
Analyze polyhistidine-tagged fusion proteins by ELISA with these high-capacity plates treated to maximize histidine-tagged protein binding.
$403.05 - $404.03
Specifications
Binding Target His tag
Quantity 5 plates
Sufficient For Binding approx. 35pmol His-tagged protein per well
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. Description Price Quantity  
Catalog Number Mfr. No. Description Price Quantity  
PI15143 Thermo Scientific™
PI15143
Pierce Copper Coated High Capacity Plates, Clear, 96-Well Pack of 5 Each for $404.03
PI15146 Thermo Scientific™
PI15146
Pierce Copper Coated High Capacity Plates, Clear, 96-Well Strip Pack of 5 Each for $403.05
Specifications
Description
Thermo Scientific™ Pierce Copper Coated High Binding Capacity (HBC) Plates use an exclusive coating process to increase the amount of histidine-tagged protein that will bind to the plate surface.
Pierce Copper Coated Plates are ideal for analyzing polyhistidine-tagged fusion proteins by ELISA-based methods. Proteins that contain a succession of several histidine residues at the amino or carboxyl terminus have a strong binding affinity for metals such as copper. These copper-coated plates are available as clear, white or black plates that can be used with colorimetric, chemiluminescent or fluorescent detection methods, respectively. Although copper is less discriminating in its binding specificity than Nickel, the higher binding capacity of these plates is ideal for high throughput screening applications that need improved sensitivity and greater dynamic range.
Highlights:
  • Quantitate previously undetectable histidine-tagged proteins
  • Wider dynamic range with four-fold greater capacity than regular nickel chelate-coated plate
  • Copper chelate provides greater binding capacity than nickel
  • Detection limit is 0.1pg of polyhistidine tagged fusion protein per well
  • Pre-blocked for the ultimate convenience
Description & Specifications
Specifications
His tag
5 plates
Binding approx. 35pmol His-tagged protein per well