Thermo Scientific™ Pierce™ High Binding Streptavidin Coated Plates
Bind biotinylated oligonucleotides and peptides with higher binding efficiency than other commercially available streptavidin-coated plates.
$329.02 - $636.32
Specifications
Binding Target Biotin
Quantity 5 plates
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. Description Price Quantity  
Catalog Number Mfr. No. Description Price Quantity  
PI15500 Thermo Scientific™
PI15500
Pierce Streptavidin Coated High Capacity Plates, Clear, 96-Well Pack of 5 Each for $329.02
PI15501 Thermo Scientific™
15501
Pierce Streptavidin Coated High Capacity Plates, Clear, 96-Well Strip Pack of 5 Each for $354.80
PI15502 Thermo Scientific™
15502
Pierce Streptavidin Coated High Capacity Plates, White, 96-Well Pack of 5 Each for $345.83
PI15503 Thermo Scientific™
15503
Pierce Streptavidin Coated High Capacity Plates, Black, 96-Well Pack of 5 Each for $345.83
PI15504 Thermo Scientific™
15504
Pierce Streptavidin Coated High Capacity Plates, Clear, 384-Well Pack of 5 Each for $564.05
PI15505 Thermo Scientific™
15505
Pierce Streptavidin Coated High Capacity Plates, White, 384-Well Each for $636.32
PI15506 Thermo Scientific™
15506
Pierce Streptavidin Coated High Capacity Plates, Black, 384-Well Pack of 5 Each for $636.32
Specifications
Description
Thermo Scientific™ Pierce Streptavidin High Binding Capacity (HBC) Coated Plates are designed for binding biotinylated oligonucleotides and peptides with higher binding efficiency than other commercially available plates.
Pierce Streptavidin High Binding Capacity (HBC) Plates are prepared with an exclusive coating method that yields a streptavidin-coated plate with four to five times the binding capacity of other commercially available polystyrene assay plates. Using a Pierce Streptavidin HBC Plate can result in an assay with a broader dynamic range and better linearity, leading to improved assay precision. The plates are offered in several plate formats, plate colors and coating volumes.
Highlights:
  • Exclusive plate-coating technology – four to five times the capacity of typical biotin-binding plates
  • Broader dynamic range – extends the quantitative range so there is no need for dilutions
  • Better sensitivity – increased binding capacity allows direct detection of small ligands not observed with regular binding capacity plates
  • Superior assay precision – standard curve demonstrates greater linearity
  • Save time – pre-blocked to reduce number of assay steps
  • Flexible assay formats – offered in 96- and 384-well formats and in different colors
Description & Specifications
Specifications
Biotin
5 plates