Thermo Scientific™ Pierce™ High Binding NeutrAvidin™ Coated Plates
Attain higher biotin-binding efficiency with these pre-blocked, ready-to-use high-capacity NeutrAvidin protein-coated microplates.
$294.84 - $603.18
Specifications
Binding Target Biotin
Quantity 5 plates
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. Description Price Quantity  
Catalog Number Mfr. No. Description Price Quantity  
PI15507 Thermo Scientific™
PI15507
Pierce NeutrAvidin Coated High Capacity Plates, Clear, 96-Well Each for $294.84
PI15508 Thermo Scientific™
PI15508
Pierce NeutrAvidin Coated High Capacity Plates, Clear, 96-Well Strip Each for $302.73
PI15509 Thermo Scientific™
PI15509
Pierce NeutrAvidin Coated High Capacity Plates, White, 96-Well Each for $320.49
PI15510 Thermo Scientific™
PI15510
Pierce NeutrAvidin Coated High Capacity Plates, Black, 96-Well Each for $322.13
PI15512 Thermo Scientific™
PI15512
Pierce NeutrAvidin Coated High Capacity Plates, White, 384-Well Each for $603.18
Specifications
Description
Thermo Scientific™ Pierce NeutrAvidin High Binding Capacity (HBC) Coated Plates are pre-blocked and ready-to-use polystyrene microplates for assays involving biotinylated antibodies or biotin-labeled molecules.
Biotin-binding plates are ideal for applications when direct adsorption to polystyrene denatures antibodies or the target molecule. Thermo Scientific NeutrAvidin™ Protein is avidin that has been deglycosylated to prevent nonspecific lectin-binding. NeutrAvidin Protein has a near-neutral pI (6.3), resulting in lower nonspecific interactions in certain applications compared with streptavidin. NeutrAvidin Protein has an additional advantage of lacking the bacterial RYD sequence found on streptavidin, which eliminates nonspecific binding due to the RGD binding domain of adhesion receptors present in a variety of cells. Pierce NeutrAvidin High Binding Capacity (HBC) Coated Plates are available in clear, white and black plates for use with colorimetric, chemiluminescent and fluorescent detection methods, respectively.
Highlights:
  • Exclusive plate-coating technology – approx. twice the capacity of typical biotin-binding plates
  • Robust and sensitive – NeutrAvidin Protein yields less nonspecific binding for improved signal-to-noise ratios
  • Broad dynamic range – extends the quantitative range so there is no need for dilutions
  • Save time – pre-blocked plates (two types available) reduces the number of assay steps
  • Flexible assay formats – choose 96- and 384-well formats and in different colors
  • NeutrAvidin Protein – performance equals or exceeds streptavidin but costs less
Description & Specifications
Specifications
Biotin
5 plates