Please login to your online account to display your discounted pricing

Thermo Scientific™ Pierce™ High Binding NeutrAvidin™ Coated Plates


Attain higher biotin-binding efficiency with these pre-blocked, ready-to-use high-capacity NeutrAvidin protein-coated microplates.

Manufacturer: thermo scientific™  PI15512

 View more versions of this product

Catalog No. PI15512

  • / Each
 

Description & Specifications

Specifications

Formulation White, 384-well plates, coated with 50uL of NeutrAvidin Protein and blocked with 100uL of SuperBlock Blocking Buffer
Quantity 5 plates
Sufficient For Binding approx. 35pmol biotin per well
Binding Target Biotin
Description Pierce NeutrAvidin Coated High Capacity Plates, White, 384-Well

Thermo Scientific™ Pierce NeutrAvidin High Binding Capacity (HBC) Coated Plates are pre-blocked and ready-to-use polystyrene microplates for assays involving biotinylated antibodies or biotin-labeled molecules.

Biotin-binding plates are ideal for applications when direct adsorption to polystyrene denatures antibodies or the target molecule. Thermo Scientific NeutrAvidin™ Protein is avidin that has been deglycosylated to prevent nonspecific lectin-binding. NeutrAvidin Protein has a near-neutral pI (6.3), resulting in lower nonspecific interactions in certain applications compared with streptavidin. NeutrAvidin Protein has an additional advantage of lacking the bacterial RYD sequence found on streptavidin, which eliminates nonspecific binding due to the RGD binding domain of adhesion receptors present in a variety of cells. Pierce NeutrAvidin High Binding Capacity (HBC) Coated Plates are available in clear, white and black plates for use with colorimetric, chemiluminescent and fluorescent detection methods, respectively.

 Highlights:

  • Exclusive plate-coating technology – approx. twice the capacity of typical biotin-binding plates

  • Robust and sensitive – NeutrAvidin Protein yields less nonspecific binding for improved signal-to-noise ratios

  • Broad dynamic range – extends the quantitative range so there is no need for dilutions

  • Save time – pre-blocked plates (two types available) reduces the number of assay steps

  • Flexible assay formats – choose 96- and 384-well formats and in different colors

  • NeutrAvidin Protein – performance equals or exceeds streptavidin but costs less