Please login to your online account to display your discounted pricing

Thermo Scientific™ Pierce™ Heptafluorobutyric Acid (HFBA)

Bottles and ampules of 99.5%-pure HFBA (MW 214.04, CAS 375-22-4), valid for use as an ion-pairing agent in reverse-phase peptide separations (HPLC).

$191.34 - $629.55

Specifications

Item Name Pierce Heptafluorobutyric Acid (HFBA)
Formulation Clear, colorless liquid of HFBA (at least 99.5% pure; zero sulfate; low water; low absorbance)
For Use With RP-HPLC
Hazards Corrosive, Irritant
Standard Components Heptafluorobutyric acid
View More Specs

 Disclaimers

Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

PI25003

 
thermo scientific™
PI25003
100mL Pack for $629.55

PI53104

 
thermo scientific™
PI53104
10 x 1mL Pack of 10 Each for $191.34
Description & Specifications

Specifications

Item Name Pierce Heptafluorobutyric Acid (HFBA)
Formulation Clear, colorless liquid of HFBA (at least 99.5% pure; zero sulfate; low water; low absorbance)
For Use With RP-HPLC
Hazards Corrosive, Irritant
Standard Components Heptafluorobutyric acid
Standard Concentration 98 to 100%
Storage Room Temperature

Thermo Scientific™ Pierce Heptafluorobutyric Acid (HFBA) is manufactured and tested to meet strict specifications that ensure superior performance for use as an ion-pairing agent in reverse-phase peptide separations.

Highlights:

  • Typical purity is 99.7% by GC; <0.1% water
  • Sequencing reagent for classical and automated Edman degradation of peptides and proteins
  • Density: 1.645
  • B.P. 120°C
  • Packaged under nitrogen in amber glass ampules or bottles
  • Clear, colorless liquid

Ordering Information:

X in the ordering table indicates that hazardous shipping charges apply.