Thermo Scientific™ Pierce™ GST Agarose
Isolate or remove cross-reacting GST-specific serum or other antibody solutions with immobilized glutathione S-transferase agarose.
$834.43 - $834.43
Specifications
Support Type Agarose
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. Description Quantity Price Quantity  
Catalog Number Mfr. No. Description Quantity Price Quantity  
PI20211 Thermo Scientific™
PI20211
Pierce GST Agarose 5mL Pack for $834.43
Specifications
Description
Perform isolation or removal of cross-reacting GST-specific antibodies from serum or other antibody solutions with Thermo Scientific™ Pierce GST Agarose.
This GST resin consists of purified glutatione S-transferase protein from Schistosoma japonicum that has been immobilized on crosslinked 6% beaded agarose. Antibody molecules in that cross-react with the GST portion of a recombinant fusion protein can be removed from the desired protein-specific antibody by passing antiserum over a GST agarose column. GST antibodies will bind to the immobilized GST, whereas antibodies against other fusion proteins will pass through the column unrestrained.
Highlights:
  • Specific – provides purification of target antibodies from those directed against the GST tag
  • Reusable – bound antibodies can be eluted with 6M guanidinium chloride allowing for 10 purifications with each column or aliquot of resin
  • Two formats – choose either pre-packed columns or resin slurry for use at a variety of scales
Description & Specifications
Specifications
Agarose