Thermo Scientific™ Pierce™ Glutathione Coated Plates
Capture and detect glutathione-S-transferase (GST) fusion proteins with these pre-blocked and glutathione (GSH)-coated microplates.
$302.64 - $324.30
Specifications
Binding Target GST tag
Quantity 5 plates
Sufficient For Binding approx. 10ng GST protein per well
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. Description Price Quantity  
Catalog Number Mfr. No. Description Price Quantity  
PI15140 Thermo Scientific™
PI15140
Pierce Glutathione Coated Plates, Clear, 96-Well Strip Pack of 5 Each for $307.90
PI15240 Thermo Scientific™
PI15240
Pierce Glutathione Coated Plates, White, 96-Well Pack of 5 Each for $302.64
PI15340 Thermo Scientific™
PI15340
Pierce Glutathione Coated Plates, Black, 96-Well Pack of 5 Each for $324.30
Specifications
Description
Thermo Scientific™ Pierce Glutathione Coated Plates are standard 96-well microplates coated with intact glutathione (GSH) and pre-blocked for use in capture and detection assays involving glutathione-S-transferase (GST) fusion proteins.
These glutathione coated plates can be used in the analysis of cell lysates to determine the presence and concentration of the GST-fusion protein produced in an expression system. Alternatively, the plates can be used to capture and coat fusion proteins via their GST tag for subsequent binding assays involving the recombinant protein (either protein interactions or a specific antibody). Because the plates are pre-blocked, initial purification of the cell lysates is not necessary.
Highlights:
  • Selective – specifically binds fusion proteins containing glutathione S-transferase
  • Capacity – binds approx. 10ng GST protein per well
  • Versatile – ready-to-bind plate format is useful for protein interaction studies
  • Compatible – detergents used to lyse cells do not inhibit GST protein binding to pre-coated plates (by contrast with direct coating methods with polystyrene plates)
  • Ready to use – pre-blocked with BSA and SuperBlock™ Blocking Buffer to reduce nonspecific binding
  • Three formats – available as clear, white or black 96-well microplates
Description & Specifications
Specifications
GST tag
5 plates
Binding approx. 10ng GST protein per well