Please login to your online account to display your discounted pricing

Thermo Scientific Pierce™ G2 Fast Blotter - Power Stainer Upgrade

Get rapid and efficient protein transfer at your fingertips with the Thermo Scientific™ Pierce™ G2 Fast Blotter - Power Stainer Upgrade.

$1,444.20 - $1,444.20

Specifications

Formulation Plastic and metal apparatus, non-powered, 25.7 x 27.8 x 3.5cm and USB memory device
For Use With (Application) Upgrade one Pierce G2 Fast Blotter Control Unit (#62291) to have electrophoretic gel staining capability
Quantity 2-part unit
View More Specs
Products
Catalog Number Mfr. No. Description For Use With (Application) Formulation Includes Price Quantity    

PI62287SPCL

 
thermo scientific
62287SPCL
Pierce G2 Fast Blotter - Protein Staining Upgrade Welcome Pack Upgrade one Pierce G2 Fast Blotter Control Unit (#62291) to have electrophoretic gel staining capability Plastic and metal apparatus, non-powered, 25.7 x 27.8 x 3.5cm and USB memory device Pierce G2 Fast Blotter - Power Stainer Upgrade (Part No. 62287), Pierce Mini Gel Power Staining Kit (Part No. 22840) Each for $1,444.20
Description & Specifications

Specifications

Formulation Plastic and metal apparatus, non-powered, 25.7 x 27.8 x 3.5cm and USB memory device
For Use With (Application) Upgrade one Pierce G2 Fast Blotter Control Unit (#62291) to have electrophoretic gel staining capability
Quantity 2-part unit

Get rapid and efficient protein transfer at your fingertips with the Thermo Scientific™ Pierce™ G2 Fast Blotter - Power Stainer Upgrade.

The Thermo Scientific Pierce G2 Fast Blotter - Power Stainer Upgrade adds Coomassie protein staining functionality to the G2 Fast Blotter.

The Pierce G2 Fast Blotter - Power Stainer Upgrade enables Pierce G2 Fast Blotter owner to update their equipment with the complete Pierce Power System software. In addition to interface updates, the upgraded G2 Fast Blotter is compatible with the Pierce Power Stain Cassette and enables rapid protein staining of SDS-PAGE gels.

For researchers only requiring semi-dry protein transfer capabilities, please see the Pierce Power Blotter.

Highlights:

  • New interface – update the G2 Fast Blotter with the new visual interface and easy-touch programming
  • New functionality – turn the G2 Fast Blotter into a Power Stainer by swapping cassettes
  • Save more time – stain SDS-PAGE protein gels in as little 6 minutes.

Includes:
  • Upgrade kit includes the Pierce Power Stain Cassette and USB Flash Drive containing the Pierce Power System upgrade software

Applications:
  • Enables the Pierce G2 Fast Blotter to be used with the Pierce Power Stain Cassette, and Pierce Power Staining Kits for rapid Coomassie protein staining in SDS-PAGE gels

Requires:
  • Pierce Power Staining Kits