Please login to your online account to display your discounted pricing

Thermo Scientific™ Pierce™ G2 Fast Blotter


Designed specifically for rapid semi-dry transfer of 10-300kDa proteins from polyacrylamide gels to nitrocellulose or PVDF membranes in 5 to 10 minutes.

Manufacturer: thermo scientific™  62291

 View more versions of this product

Catalog No. PI62291

  • / Each
 

Description & Specifications

Specifications

Quantity 1 unit
Formulation Power supply with touchscreen user interface, 16.4 x 27.6 x 15.4cm
Kit Contents Pierce G2 Fast Blotter Control Unit
Description Pierce G2 Fast Blotter Control Unit
Sufficient For Semi-dry transfer when used with Pierce G2 Fast Blotter Cassette

The Thermo Scientific™ Pierce G2 Fast Blotter is designed specifically for rapid semi-dry transfer of 10-300kDa proteins from polyacrylamide gels to nitrocellulose or PVDF membranes in 5 to 10 minutes.

The Pierce G2 Fast Blotter features an integrated power supply optimized to deliver consistent, high-efficiency protein transfer with commonly used pre-cast and homemade gels (SDS-PAGE) and nitrocellulose or PVDF membranes. The large cassette provides for simultaneous transfer of up to four mini-sized gels or two midi-sized gels. The blotter has an easy-to-use color LCD touchscreen interface and pre-programmed transfer methods for different numbers and sizes of gels and molecular weight ranges of proteins. The easy-touch programming feature allows custom transfer settings to be quickly created, saved and run.
The Pierce G2 Fast Blotter achieves highly efficient transfer in less than 10 minutes when used with Pierce 1-Step Transfer Buffer (Part No. 84731). The instrument is also effective for standard 30- to 60-minute semi-dry transfer protocols based on traditional buffers.

Highlights:

  • Fast - transfer proteins in 5 to 10 minutes when used with 1-Step Transfer Buffer
  • Efficient - achieve high transfer efficiency with a broad range of protein sizes (10-300kDa) compared to conventional semi-dry or wet (tank) transfer methods
  • Integrated power supply - seamless operation between control unit and cassette provides consistent high efficiency protein transfer
  • Easy-touch programming - access pre-programmed transfer methods or create, save and run customized transfer methods
  • Convenient - simultaneously transfer 1 to 4 mini gels or 1 to 2 medium-sized gels
  • Versatile - use with Pierce 1-Step Transfer Buffer for rapid blotting programs or Towbin transfer buffer for conventional semi-dry transfer methods

Applications: Semi-dry Western blot transfer of 10-300kDa proteins from polyacrylamide gels to nitrocellulose or PVDF membranes

Includes: System consists of the Pierce G2 Fast Blotter Control Unit and Pierce G2 Fast Blotter Cassette; kit includes Western Blot Roller, Power Cord with C/13 Connector, and Quick Start Guide. The unit ships with North American power cord; also available are country-specific power cords.