Thermo Scientific™ Pierce™ Ethidium Bromide
Visualize DNA fluorescently in a variety of molecular biology applications using these dilute (0.625mg/mL) or concentrated (10mg/mL) ethidium bromide solutions.
$86.87 - $291.34
Specifications
For Use With (Application) DNA fluorescent visualization
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
PI17896 Thermo Scientific™
PI17896
5mL Pack for $86.87
PI17898 Thermo Scientific™
PI17898
5 x 10mL Pack of 5 Each for $291.34
Specifications
Description
Thermo Scientific™ Pierce Ethidium Bromide is available as both dilute (0.625mg/mL) and more concentrated (10mg/mL) solutions to accommodate most molecular biology applications for fluorescent DNA visualization.
Thermo Scientific™ Pierce Ethidium Bromide is available as both dilute (0.625mg/mL) and more concentrated (10mg/mL) solutions to accommodate most molecular biology applications for fluorescent DNA visualization.
Highlights:
0.625mg/mL EtBr Solution:
  • Each drop contains 25μg of ethidium bromide and is ideal for a 50mL agarose gel
  • After adding one drop of solution per 50mL of gel, the final concentration of EtBr will be 0.5μg/mL, which is the recommended concentration for electrophoresis of nucleic acids
  • Can also be used as the running buffer or for staining gels after electrophoresis
10mg/mL EtBr Solution:
  • A safe and convenient way to use ethidium bromide while minimizing exposure
Description & Specifications
Specifications
DNA fluorescent visualization