Thermo Scientific™ Pierce™ EGFR In-Cell ELISA Kit
Measure total and phosphorylated epidermal growth factor receptor (phospho-EGFR) using EGFR antibodies detected by fluorescence or TMB substrate in whole-cell plate assays.
Manufacturer:  Thermo Scientific™ 62205
 View more versions of this product
Catalog No. PI62205
Description
Description
Thermo Scientific™ Pierce EGFR In-Cell ELISA Kits are colorimetric and fluorescent microplate assays optimized to measure total and phosphorylated epidermal growth factor receptor (phospho-EGFR) in whole cells.
The protein encoded by this gene is a transmembrane glycoprotein that is a member of the protein kinase superfamily. This protein is a receptor for members of the epidermal growth factor family. EGFR is a cell surface protein that binds to epidermal growth factor. Binding of the protein to a ligand induces receptor dimerization and tyrosine autophosphorylation and leads to cell proliferation. Mutations in this gene are associated with lung cancer. Multiple alternatively spliced transcript variants that encode different protein isoforms have been found for this gene.
Specifications
Specifications
P00533
Pierce EGFR Colorimetric In-Cell ELISA Kit
1956
Blocking Buffer, Components included only in the target-specific kits:Antibody #1 and Antibody #2, Elution Buffer, 2 x 30mL, 20X Tris Buffered Saline, Surfact-Amps 20, Surfact-Amps X-100, HRP Conjugate, TMB Substrate, TMB Stop Solution, Janus Green Whole-Cell Stain, Elution Buffer, Thin Plate Seal Assembly
96 test kit
Supernatants
2-8°C
1 hr 20 min
Human
HRP
HRP
Multi-component kit with HRP-conjugate
Colorimetric Microplate Reader
ELISA
RUO
Supernatants (10,000 cells)
96 microplate wells of overnight-cultured cells
24 hr
EGFR
Documents