Please login to your online account to display your discounted pricing

Thermo Scientific™ Pierce™ DTT (Dithiothreitol)


Reduce disulfides in biological buffers and sample loading buffers with pure, crystalline dithiothreitol (DTT), the most popular protein disulfide reducing agent.

Manufacturer: thermo scientific™  PI20291

 View more versions of this product

Catalog No. PI20291

  • / Pack
 

Description & Specifications

Specifications

Description Pierce DTT, No-Weigh Format
Formulation

Pure (approx. 98%) white powder of dithiothreitol

Quantity 48 x 7.7mg
Sufficient For Preparing 100uL of 0.5M DTT per microtube

Thermo Scientific™ Pierce DTT (Dithiothreitol) is offered as 7.7mg × 48-tube microplates for convenient preparation of 100μL of 0.5M stocks for addition to protein samples for effective reduction of disulfide bonds.

Highlights:

  • A water-soluble reagent that reduces disulfide bonds
  • Maintains mono-thiols completely in the reduced state and reduces disulfide bonds quantitatively
  • Specific and sensitive assay for disulfides using DTT with Ellman's Reagent