Thermo Scientific™ Pierce™ Disposable Plastic Columns
Empty drip columns (0.5 to 5 mL) with matching frits and caps for packing beaded affinity or desalting resins to use in gravity-flow purification techniques.
$116.38 - $345.71
Products ${productFamilyLength}
Catalog Number Mfr. No. Description Price Quantity  
Catalog Number Mfr. No. Description Price Quantity  
PI29920 Thermo Scientific™
PI29920
Disposable Columns, 2mL Pack of 100 Each for $279.66
PI29922 Thermo Scientific™
PI29922
Disposable Columns, 5mL Pack of 100 Each for $322.52
PI29923 Thermo Scientific™
PI29923
Disposable Column Funnels Pack of 50 Each for $212.94
PI29924 Thermo Scientific™
PI29924
Disposable Columns, 10mL Pack of 100 Each for $345.71
PI29925 Thermo Scientific™
PI29925
Disposable Column Trial Pack Pack for $116.38
Specifications
Description
Thermo Scientific™ Pierce Disposable Plastic Columns are empty columns with matching frits and caps for use in a variety of resin-based affinity or desalting techniques.
Additional Information and Online Ordering »
These bulk packages of empty columns are economical and convenient devices for packing affinity purification supports and desalting resins for gravity-flow procedures. Three columns sizes are offered, suitable for procedures requiring 0.5 to 10mL resin-bed volumes. The plastic columns are supplied with matching pairs of polyethylene discs and polypropylene caps (top and bottom). The columns are economically-priced for single use and sufficiently durable to prepare and store columns for multiple uses.
Highlights:
  • For bench top gravity-flow affinity purification and size exclusion chromatography
  • Supplied complete with porous polyethylene discs, stoppers and end caps
  • Columns can be pre-packed and stored until needed
  • Compatible with most types of aqueous buffer eluents commonly used in chromatography
  • Plastic funnels can be attached to expand the buffer reservoir for increased flow rates
Includes:
Trial pack contains two columns of each available size (six columns total), matching column accessories (discs and caps), and one funnel (column reservoir extender)